Assay Details
| Gene Symbol: pes-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001028235.3 | NP_001023406.1 | 4 - 5 | 488 | 92 | |
| NM_001383089.1 | 4 - 5 | 491 | 92 | |||
| GenBank mRNA | Z28376.1 | - | 387 | 92 | ||
| Z28377.1 | - | 484 | 92 | |||
More Information
Set Membership: |
|

pes-1
Genomic Map