Assay Details
Gene Symbol: flp-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_069191.7 | NP_501592.1 | 2 - 3 | 179 | 91 | |
NM_182277.8 | NP_872077.1 | 2 - 3 | 166 | 91 | ||
NM_182278.7 | NP_872078.1 | 2 - 3 | 158 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
