Assay Details
| Gene Symbol: asp-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_182329.5 | NP_872129.1 | 1 - 2 | 362 | 63 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

asp-2| Gene Symbol: asp-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_182329.5 | NP_872129.1 | 1 - 2 | 362 | 63 | |
Set Membership: |
|