Assay Details
| Gene Symbol: sri-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026253.3 | NP_001021424.1 | 1 - 2 | 192 | 165 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

sri-5| Gene Symbol: sri-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026253.3 | NP_001021424.1 | 1 - 2 | 192 | 165 | |
Set Membership: |
|