Assay Details
| Gene Symbol: nfya-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001264657.2 | NP_001251586.1 | 4 - 5 | 283 | 64 | |
| NM_001264658.3 | NP_001251587.1 | 3 - 4 | 206 | 64 | ||
More Information
Set Membership: |
|

nfya-2
Genomic Map