Assay Details
| Gene Symbol: dys-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001306310.3 | NP_001293239.1 | 33 - 34 | 8864 | 111 | |
| NM_001306311.3 | NP_001293240.1 | 22 - 23 | 6065 | 111 | ||
| NM_001306312.3 | NP_001293241.1 | 10 - 11 | 3251 | 111 | ||
| NM_001306313.4 | NP_001293242.1 | 36 - 37 | 9509 | 111 | ||
| NM_001306314.3 | NP_001293243.1 | 32 - 33 | 8831 | 111 | ||
| NM_001383731.1 | 32 - 33 | 8882 | 111 | |||
| NM_060545.6 | NP_492946.1 | 37 - 38 | 9604 | 111 | ||
More Information
Set Membership: |
|

dys-1
Genomic Map