Assay Details
Gene Symbol: spe-9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_060554.5 | NP_492955.1 | 1 - 2 | 70 | 135 | |
GenBank mRNA | AF049327.1 | - | 1 - 2 | 48 | 135 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
