Assay Details
Gene Symbol: spe-9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_060554.5 | NP_492955.1 | 6 - 7 | 719 | 88 | |
GenBank mRNA | AF049327.1 | - | 6 - 7 | 697 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
