Assay Details
Gene Symbol: spe-9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_060554.5 | NP_492955.1 | 8 - 9 | 891 | 85 | |
GenBank mRNA | AF049327.1 | - | 8 - 9 | 869 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
