Assay Details
| Gene Symbol: ubc-12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_060623.6 | NP_493024.1 | 1 - 2 | 97 | 63 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ubc-12| Gene Symbol: ubc-12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_060623.6 | NP_493024.1 | 1 - 2 | 97 | 63 | |
Set Membership: |
|