Assay Details
| Gene Symbol: hda-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_060625.7 | NP_493026.1 | 3 - 4 | 846 | 83 | |
 Back To Top
 Back To TopMore Information
| Set Membership: |  Amplicon length between 71 and 85  Recommended Assay  Caenorhabditis elegans  Probe spans exons | 
Gene Ontology Categories:
 Back To Top
 Back To Top
 hda-3
hda-3 Genomic Map
Genomic Map Back To Top
 Back To Top