Assay Details
Gene Symbol: clp-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_060651.7 | NP_493052.3 | 8 - 9 | 1500 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
