Assay Details
Gene Symbol: dlk-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026272.4 | NP_001021443.1 | 3 - 4 | 370 | 122 | |
NM_001026273.6 | NP_001021444.1 | 3 - 4 | 370 | 122 | ||
NM_001026274.5 | NP_001021445.1 | 3 - 4 | 365 | 122 | ||
NM_001136297.4 | NP_001129769.1 | 1 - 2 | 146 | 122 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
