Assay Details
Gene Symbol: dlk-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026272.4 | NP_001021443.1 | 9 - 10 | 2049 | 62 | |
NM_001136297.4 | NP_001129769.1 | 7 - 8 | 1825 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
