Assay Details
Gene Symbol: sra-18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_060813.1 | NP_493214.1 | 3 - 4 | 662 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
