Assay Details
| Gene Symbol: sra-22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_060817.2 | NP_493218.2 | 4 - 5 | 869 | 131 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

sra-22| Gene Symbol: sra-22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_060817.2 | NP_493218.2 | 4 - 5 | 869 | 131 | |
Set Membership: |
|