Assay Details
| Gene Symbol: sra-24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_060819.2 | NP_493220.2 | 1 - 2 | 256 | 113 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

sra-24| Gene Symbol: sra-24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_060819.2 | NP_493220.2 | 1 - 2 | 256 | 113 | |
Set Membership: |
|