Assay Details
| Gene Symbol: smu-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_060878.6 | NP_493279.1 | 3 - 4 | 325 | 139 | |
| GenBank mRNA | AF330595.1 | - | 3 - 4 | 297 | 139 | |
More Information
Set Membership: |
|

smu-1
Genomic Map