Assay Details
| Gene Symbol: smu-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_060878.6 | NP_493279.1 | 4 - 5 | 724 | 69 | |
| GenBank mRNA | AF330595.1 | - | 4 - 5 | 696 | 69 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

smu-1| Gene Symbol: smu-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_060878.6 | NP_493279.1 | 4 - 5 | 724 | 69 | |
| GenBank mRNA | AF330595.1 | - | 4 - 5 | 696 | 69 | |
Set Membership: |
|