Assay Details
| Gene Symbol: smu-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_060878.6 | NP_493279.1 | 8 - 9 | 1366 | 68 | |
| GenBank mRNA | AF330595.1 | - | 8 - 9 | 1338 | 68 | |
More Information
Set Membership: |
|

smu-1
Genomic Map