Assay Details
Gene Symbol: pals-12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047172.1 | NP_001040637.1 | 3 - 4 | 839 | 78 |
Gene Symbol: pals-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_132565.1 | 7 - 8 | 1709 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
