Assay Details
Gene Symbol: F22G12.5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001265159.4 | NP_001252088.2 | 20 - 21 | 4447 | 76 | |
NM_001265160.3 | NP_001252089.1 | 6 - 7 | 1233 | 76 | ||
NM_001313464.2 | NP_001300393.1 | 20 - 21 | 4453 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
