Assay Details
Gene Symbol: top-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383741.1 | 3 - 4 | 2236 | 75 | ||
NM_060936.7 | NP_493337.1 | 4 - 5 | 2455 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
