Assay Details
Gene Symbol: spr-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_060965.6 | NP_493366.1 | 4 - 5 | 1265 | 119 | |
GenBank mRNA | AX799789.1 | - | 4 - 5 | 1263 | 119 | |
AY152852.1 | - | 4 - 5 | 1263 | 119 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
