Assay Details
| Gene Symbol: trp-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_061028.3 | NP_493429.2 | 14 - 15 | 3458 | 53 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

trp-4| Gene Symbol: trp-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_061028.3 | NP_493429.2 | 14 - 15 | 3458 | 53 | |
Set Membership: |
|