Assay Details
| Gene Symbol: riok-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_061143.8 | NP_493544.2 | 1 - 2 | 202 | 67 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

riok-2| Gene Symbol: riok-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_061143.8 | NP_493544.2 | 1 - 2 | 202 | 67 | |
Set Membership: |
|