Assay Details
| Gene Symbol: tep-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001381276.2 | NP_001366904.1 | 12 - 13 | 2644 | 112 | |
| NM_001381277.1 | NP_001367060.1 | 13 - 14 | 2624 | 112 | ||
| NM_061213.5 | NP_493614.1 | 13 - 14 | 2595 | 112 | ||
More Information
Set Membership: |
|

tep-1
Genomic Map