Assay Details
Gene Symbol: rpt-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026942.5 | NP_001022113.1 | 3 - 4 | 1183 | 115 | |
NM_001026943.4 | NP_001022114.1 | 2 - 3 | 1078 | 115 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
