Assay Details
Gene Symbol: etr-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001381290.2 | NP_001367867.1 | 7 - 8 | 1177 | 112 | |
NM_001381302.1 | NP_001367869.1 | 3 - 4 | 419 | 112 | ||
NR_132573.1 | 7 - 8 | 1115 | 112 | |||
GenBank mRNA | EF523766.1 | - | 7 - 8 | 1115 | 112 | |
U53931.1 | - | 7 - 8 | 1178 | 112 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
