Assay Details
| Gene Symbol: chs-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001313615.3 | NP_001300544.1 | 10 - 11 | 2674 | 129 | |
| NM_001313616.3 | NP_001300545.1 | 12 - 13 | 2956 | 129 | ||
| NM_061281.6 | NP_493682.2 | 10 - 11 | 3084 | 129 | ||
| GenBank mRNA | AY874872.1 | - | 10 - 11 | 2860 | 129 | |
More Information
Set Membership: |
|

chs-2
Genomic Map