Assay Details
| Gene Symbol: chs-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001313615.3 | NP_001300544.1 | 16 - 17 | 4633 | 64 | |
| NM_001313616.3 | NP_001300545.1 | 18 - 19 | 4915 | 64 | ||
| NM_061281.6 | NP_493682.2 | 16 - 17 | 5043 | 64 | ||
| GenBank mRNA | AY874872.1 | - | 16 - 17 | 4819 | 64 | |
More Information
Set Membership: |
|

chs-2
Genomic Map