Assay Details
| Gene Symbol: nfyb-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001316872.3 | NP_001303801.1 | 6 - 7 | 800 | 64 | |
| NM_061339.6 | NP_493740.1 | 7 - 8 | 1051 | 64 | ||
More Information
Set Membership: |
|

nfyb-1
Genomic Map