Assay Details
| Gene Symbol: mlt-8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_061358.8 | NP_493759.2 | 5 - 6 | 3620 | 78 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

mlt-8| Gene Symbol: mlt-8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_061358.8 | NP_493759.2 | 5 - 6 | 3620 | 78 | |
Set Membership: |
|