Assay Details
| Gene Symbol: pho-14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_061510.3 | NP_493911.1 | 9 - 10 | 1216 | 81 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

pho-14| Gene Symbol: pho-14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_061510.3 | NP_493911.1 | 9 - 10 | 1216 | 81 | |
Set Membership: |
|