Assay Details
| Gene Symbol: decr-1.1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_061850.4 | NP_494251.1 | 1 - 2 | 148 | 80 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

decr-1.1| Gene Symbol: decr-1.1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_061850.4 | NP_494251.1 | 1 - 2 | 148 | 80 | |
Set Membership: |
|