Assay Details
| Gene Symbol: vab-19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_061875.3 | NP_494276.2 | 11 - 12 | 2203 | 62 | |
| GenBank mRNA | AY273823.1 | - | 11 - 12 | 2190 | 62 | |
More Information
Set Membership: |
|

vab-19
Genomic Map