Assay Details
Gene Symbol: K02F6.4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_061892.1 | NP_494293.1 | 4 - 5 | 2606 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: K02F6.4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_061892.1 | NP_494293.1 | 4 - 5 | 2606 | 67 |
Set Membership: |
![]() ![]() ![]() ![]() |