Assay Details
| Gene Symbol: F22E5.17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_061929.7 | NP_494330.2 | 1 - 2 | 120 | 71 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

F22E5.17| Gene Symbol: F22E5.17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_061929.7 | NP_494330.2 | 1 - 2 | 120 | 71 | |
Set Membership: |
|