Assay Details
| Gene Symbol: pigw-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027236.3 | NP_001022407.2 | 4 - 5 | 1026 | 62 | |
| NM_001381355.1 | NP_001367965.1 | 2 - 3 | 250 | 62 | ||
More Information
Set Membership: |
|

pigw-1
Genomic Map