Assay Details
| Gene Symbol: pigw-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027236.3 | NP_001022407.2 | 5 - 6 | 1170 | 68 | |
| NM_001381355.1 | NP_001367965.1 | 3 - 4 | 394 | 68 | ||
More Information
Set Membership: |
|

pigw-1
Genomic Map