Assay Details
| Gene Symbol: nra-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_070966.4 | NP_503367.1 | 7 - 8 | 3273 | 76 | |
| NM_182391.2 | NP_872191.1 | 7 - 8 | 3281 | 76 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

nra-4| Gene Symbol: nra-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_070966.4 | NP_503367.1 | 7 - 8 | 3273 | 76 | |
| NM_182391.2 | NP_872191.1 | 7 - 8 | 3281 | 76 | ||
Set Membership: |
|