Assay Details
| Gene Symbol: cblc-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027353.7 | NP_001022524.1 | 3 - 4 | 556 | 111 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

cblc-1| Gene Symbol: cblc-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027353.7 | NP_001022524.1 | 3 - 4 | 556 | 111 | |
Set Membership: |
|