Assay Details
| Gene Symbol: rpl-22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_062531.7 | NP_494932.1 | 2 - 3 | 247 | 94 | |
| NR_132580.1 | 2 - 3 | 319 | 94 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

rpl-22| Gene Symbol: rpl-22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_062531.7 | NP_494932.1 | 2 - 3 | 247 | 94 | |
| NR_132580.1 | 2 - 3 | 319 | 94 | |||
Set Membership: |
|