Assay Details
| Gene Symbol: gcy-12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_062594.3 | NP_494995.2 | 6 - 7 | 1416 | 99 | |
| NR_134556.1 | 6 - 7 | 1342 | 99 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

gcy-12| Gene Symbol: gcy-12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_062594.3 | NP_494995.2 | 6 - 7 | 1416 | 99 | |
| NR_134556.1 | 6 - 7 | 1342 | 99 | |||
Set Membership: |
|