Assay Details
| Gene Symbol: daz-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_062635.5 | NP_495036.3 | 5 - 6 | 1385 | 82 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

daz-1| Gene Symbol: daz-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_062635.5 | NP_495036.3 | 5 - 6 | 1385 | 82 | |
Set Membership: |
|