Assay Details
| Gene Symbol: ttc-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_062686.5 | NP_495087.1 | 1 - 2 | 122 | 86 | |
| Gene Symbol: C17G10.1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_062687.8 | NP_495088.1 | 1 - 2 | 93 | 86 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_062687.8 transcripts.
Set Membership: |
|

ttc-4
Genomic Map