Assay Details
Gene Symbol: sam-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001381430.2 | NP_001367525.1 | 4 - 5 | 776 | 76 |
Gene Symbol: F59E12.17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383848.1 | 4 - 5 | 776 | 76 |
Gene Symbol: ddl-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383847.1 | 4 - 5 | 780 | 76 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001383847.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
