Assay Details
| Gene Symbol: ehs-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027328.6 | NP_001022499.2 | 3 - 4 | 373 | 114 | |
| NR_151580.1 | 3 - 4 | 358 | 114 | |||
| GenBank mRNA | AY027560.1 | - | 3 - 4 | 380 | 114 | |
More Information
Set Membership: |
|

ehs-1
Genomic Map