Assay Details
| Gene Symbol: H41C03.1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_062767.5 | NP_495168.1 | 5 - 6 | 1049 | 100 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

H41C03.1| Gene Symbol: H41C03.1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_062767.5 | NP_495168.1 | 5 - 6 | 1049 | 100 | |
Set Membership: |
|