Assay Details
| Gene Symbol: gldc-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001348634.3 | NP_001335514.1 | 1 - 2 | 253 | 80 | |
| NM_062808.8 | NP_495209.1 | 2 - 3 | 553 | 80 | ||
More Information
Set Membership: |
|

gldc-1
Genomic Map